Liquid-Binder™ Aqueous Additive, Chemplex®
Supplier: CHEMPLEX INDUSTRIES
800
15566-086EA 320.45 USD
15566-086
Liquid-Binder™ Aqueous Additive, Chemplex®
Stains Microscopy and staining reagents
Converts powdered samples into a slurry to eliminate segregation by size or density during comminution process.
Particularly useful for compositionally complex powders consisting of particles of different size, shape, density and hardness. Ideal for XRD analysis where Liquid-Binder™ is useful in forming self-supporting cylindrical samples for direct mounting in a camera. May also be used as a dispersing/mounting medium.