Boric Acid Tablets, Chemplex
Supplier: CHEMPLEX INDUSTRIES
750
15566-084PK 158.66 USD
15566-084
Boric Acid Tablets, Chemplex
Media for Microbiology Additives and Supplements for Microbiology
Very abrasive heat-resistant additive suitable for processing samples that tend to reweld during comminution in metallic grinding vessels.
Frequently used in conjunction with x-ray mix to impart lubricity for “plasticizing” pellets.