SPILFYTER® KOLOR-LOCK® Solidifying Neutralizer for HF Acids, NPS
Supplier: NPS
472101
15552-226CS 1296.19 USD
15552-226
SPILFYTER® KOLOR-LOCK® Solidifying Neutralizer for HF Acids, NPS
Spill Control Materials
KOLOR-LOCK® super-absorbent polymers solidify the spill as it neutralizes
KOLORLOCK® takes the guesswork out of knowing when an acid is neutralized through its 3-color change indicator process.