SPILFYTER® Mercsorb® Mercury Spill Kit, Small, NPS
Supplier: NPS
520260
15552-190CS 271.21 USD
15552-190
SPILFYTER® Mercsorb® Mercury Spill Kit, Small, NPS
Spill Control Materials
Kit is useful for clean-up of small mercury spills and converts mercury into an amalgam
  • Converts small mercury spills into an amalgam
  • For use by trained personnel only
Contents of the kit includes 500 g Amalgamation Powder, 2 Mercury Sponges and 1 Containment Jar.
For use by trained personnel only.