Flux and Liquid Dispensers, Plato™, ITW Chemtronics
Supplier: ITW Chemtronics
FD-21
15551-500EA 13 USD
15551-500
Flux and Liquid Dispensers, Plato™, ITW Chemtronics
Bottles Dropping and Dispensing Bottles
POS–A–LOC® leakproof liquid dispensers with needles conveniently dispense specific amounts of a variety of liquids without dripping, spilling, or waste.
Stainless steel replacement needles also available.
The cut-to-size flux dispenser allows you to cut the tip opening to the correct size.