Two State Wrist Strap Tester, Desco Industries
Supplier: Desco
19350
15551-262EA 124.13 USD
15551-262
Two State Wrist Strap Tester, Desco Industries
Static Control Monitors
Two State Wrist Strap Tester allows for tests to be performed during normal use at workstation.
  • Connects to utility ground when plugged in
  • Tests entire circuit; wrist strap, cord, and skin contact
  • Acts as common point ground for the worksurface
  • Includes adapter
  • Range: 750 Kilohms – 10 Megohms
  • NIST calibrated
  • Made in the United States of America
Connects to utility ground when plugged in for quick and easy installation and grounding of both wrist straps and table mats. Acts as common point ground for the worksurface, with no other ground cords needed. Test can be performed during normal use at workstation to improve productivity. Provided with calibration to NIST traceable standards. Includes adapter (also available separately) if replacement is required.