VWR® Traceable® Precision Dial Barometer
Supplier: VWR International
15551-024
15551-024EA 283.61 USD
15551-024
VWR® Traceable® Precision Dial Barometer
Barometers
Triple scales provide a sensitive measure of atmospheric pressure.
  • Rugged, corrosion-proof nickel-chrome case
  • Sensitive measuring of atmospheric pressure
  • Manual pointer
  • Precise measurement of barometic changes
Unit is ideal for monitoring labs, plants, and weather changes.
Housed in a rugged, corrosion-proof nickel-chrome case. Manual pointer indicates barometric changes since last viewing. Screw adjustment compensates for altitude.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Includes notched desk stand, wall mount, and traceability certificate.