MONOSPOT® Latex Test Kit, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
776150
15205-015PK 659.44 USD
15205-015
MONOSPOT® Latex Test Kit, Meridian Bioscience
Clinical Diagnostic Supplies Clinical Reagents and Kits
Rapid latex agglutination test for the qualitative or semi-quantitative detection of Infectious Mononucleosis (IM) heterophile antibodies in human serum or plasma
Includes test cards, latex reagent, positive control, negative control, and pipet stirrers. Kit contains sufficient materials for 50 determinations. CE marked.