May Grunwald's eosin-methylene blue solution, HARLECO®, Sigma-Aldrich®
Supplier: MilliporeSigma
660-85
CA15204-200CS 2276.34 USD
CA15204-200 CA15204-292 15204-292 15204-200
May Grunwald's eosin-methylene blue solution, HARLECO®, Sigma-Aldrich®
May Grunwald's eosin-methylene blue
HARLECO® Blood stain producing results similar to Wright Stain. May be used in conjunction with Giemsa Stain for bone marrow preparations
  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use
Blood stain producing results similar to Wright Stain. May be used in conjunction with Giemsa Stain for bone marrow preparations.