Carbol fuchsin solution (Kinyoun’s), HARLECO® stain for bacteriology, for AFB (acid fast bacteria), Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 4-[(4-Amino-m-tolyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl]-o-toluidine hydrochloride

6033X-75
15204-262EA 482 USD
15204-262
Carbol fuchsin solution (Kinyoun’s), HARLECO® stain for bacteriology, for AFB (acid fast bacteria), Sigma-Aldrich®
Carbol fuchsin

HARLECO® Formulated specifically for Kinyoun stain technique for acid-fast staining (AFB)


  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use


Does not contain Tergitol®. Addition of heat is optional.

Formula: C₂₁H₂₂ClN₃
MW: 351.88 g/mol
Flash Pt: 48.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00013129
CAS Number: 4197-24-4
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR