EA Solution Gill modified (with Eosin and Light Green), HARLECO® stain for cytology, Sigma-Aldrich®
Supplier: MilliporeSigma
Danger
65068-85
15204-184EA 397 USD
15204-184
EA Solution Gill modified (with Eosin and Light Green), HARLECO® stain for cytology, Sigma-Aldrich®
EA Solution Gill
HARLECO® EA stain modified to exclude Bismarck Brown
Prepared from dyes certified by the Biological Stain Commission (BSC)