Levy Counting Chamber, Hausser Scientific
Supplier: Hausser
3500
15170-208EA 438.22 USD
15170-208
Levy Counting Chamber, Hausser Scientific
Slides Counting Chambers
Hemacytometer for accurate red and white blood cell counts
Improved double Neubauer rulings with dark lines on a clear background. Cell depth: 0.1mm. One-piece construction.
Ordering information: Counting chamber furnished with two octagonal cover glasses. Without case.