Paraplast® embedding agent for histology
Supplier: Leica Biosystems
For use as tissue embedding media.
- Paraffin and plastic polymers construction
- Low viscosity permits complete infiltration
- Suitable for clear, smooth tissue specimens
Paraplast®, made of paraffin and plastic polymers, allows clean cuts and minimizes wrinkles for clear, smooth tissue specimens
Compatible with most solvents and staining techniques. Melting point: 56°C (133°F) (ASTM).
Packed in 1kg (2.2lbs.) bags of pellets.
Packaging: Packed in 1kg (2.2lbs.) bags of pellets.
CAS Number:
8002-74-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...