Macro-CON® Concentration System, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
970120
15157-028CS 671.92 USD
15157-028
Macro-CON® Concentration System, Meridian Bioscience
Clinical Diagnostic Supplies Clinical Reagents and Kits
A closed system for the concentration of eggs, larvae, and protozoa from preserved fecal specimens
The filtration unit is used in conjunction with the Para-Pak® specimen collection vial, creating a completely closed system and minimizing user exposure to potentially infectious agents. System uses the entire contents of the collection vial, reducing variation due to specimen sampling.