CON-Trate® Concentration System, Meridian Bioscience
Supplier: MERIDIAN DIAGNOSTICS
960200
15157-026CS 1220.44 USD
15157-026
CON-Trate® Concentration System, Meridian Bioscience
Clinical Diagnostic Supplies Clinical Reagents and Kits
Standardized concentration system for the recovery of eggs, larvae, and protozoa from fecal material
System contains enough components for 200 tests. Components include filters, 15mL centrifuge tubes, caps, Reagent A (MucoPenX), and Reagent B (Ethyl Acetate).