VWR® Micro Slide Staining Rack
Supplier: VWR International
80863300
15153-821EA 128.48 USD
15153-821
VWR® Micro Slide Staining Rack
Staining Boxes
This stainless steel staining rack is designed for use across laboratory sinks of various sizes.
  • Stainless steel construction
  • Quickly adjustable
Triangular slide supporting rods have a tapered top, and contact the slides only in a narrow zone. Rack is simply and quickly adjustable to fit sinks up to 53.4 cm (21") inside dimensions.
Ordering information: Blue tray (as shown in picture) is not included with Micro Slide Staining Rack.