Microtome Knives, Leica® Microsystems
Supplier: Leica Biosystems MS
14021607100
15148-676EA 1610.78 USD
15148-676
Microtome Knives, Leica® Microsystems
Microtomes and Microtome Assemblies Microtome Blades and Handles
For biological and industrial applications requiring paraffin sectioning, cryosectioning, or soft plastic sectioning.
  • Steel blades
  • Profile options available
C-profile blades feature a V-shape bevel, useful for any method of sectioning.
Ordering information: Knives come with a case.