Soder-Wick® Desoldering Braids, ITW Chemtronics®
Supplier: ITW Chemtronics
60-5-5 60-4-10 60-4-5 60-2-10 60-2-5 60-1-10 60-1-5 60-3-10 80-2-10 80-2-5 80-1-10 80-1-5 80-6-5 80-5-10 80-4-5 80-3-5 80-3-10 50-1-25
15145-620PK 7.06 USD
15145-620 15145-622 15145-626 15145-628 15145-630 15145-634 15145-636 15145-638 15145-644 15145-646 15145-648 15145-650 15145-652 15145-654 15145-656 15145-658 15145-660 15145-700 62111-306 62111-308
Soder-Wick® Desoldering Braids, ITW Chemtronics®
Soldering Supplies Desoldering Braids
Desoldering braids significantly reduce rework/repair time and minimize the risk of heat damage
Soder-Wick® Rosin is a Type R flux braid that is fast, safe, noncorrosive, and leaves no ionic contamination.
Soder-Wick® No Clean SD braid is noncorrosive, halide-free, and organic.
Meet Bellcore TR-NWT-000078 and IPC SF-818, and ANSI/IPC J STD-004, Type ROL0.
The weave design allows maximum solder capacity and optimizes heat transfer. Braids leave little or no flux residue, shortening or completely eliminating cleaning time.
Ordering information: Soder-Wick® Rosin flux is available in either ESD-safe static-dissipative bobbins sealed in Performance Pak™ barrier packaging (25 bobbins per pack) or in individual standard spool packaging. Soder-Wick® No Clean SD flux is available on static-dissipative bobbins that are sealed in Performance Pak™ barrier packaging (25 bobbins per pack).