Coventry™ Pillow Tipped Swabs, ITW Chemtronics®
Supplier: ITW Chemtronics
52121 51121
15145-603CS 727.7 USD
15145-603 15145-605
Coventry™ Pillow Tipped Swabs, ITW Chemtronics®
Swabs Swabs for Cleaning
Pillow-tip™ swabs consist of polyester knit fabric which is held in place using a shrink tube
  • Engineered for high solvent retention with the cleanliness of ISO Class 4 processed polyester
  • Seamless design is lint-free
  • High-strength, adhesive-free swab, durable head construction
  • Excellent solvent compatibility
  • Available Aqua-Prime™ processed
Suitable for cleaning high end disk drive components, Aerospace Industry and Micro Mechanical cleaning, and to validate cleanliness of manufacturing surfaces for the Pharmaceutical and Biotechnical Industry.
These swabs are low in nonvolatile residue, ionic and particulate contamination, while providing exceptional strength and solvent resistance. Glass filled nylon handles are rigid like wooden handles but are cleanroom grade.