Ultrajet® All-Way Duster, ITW Chemtronics®
Supplier: ITW Chemtronics
ES1620
15145-526CS 457.76 USD
15145-526
Ultrajet® All-Way Duster, ITW Chemtronics®
Sprays
Multi-directional, high-pressure compressed gas duster cleans electronics without scratching delicate surfaces
Contains 100% HFC-134a filtered to 0.2 microns. Safe on plastics. Can be used on laboratory instruments, medical devices, electronics, and more. Ozone-safe. Nonflammable, CFC/HCFC-free, and residue-free.