Static Free™ Hand Guard® Lotion, ITW Chemtronics
Supplier: ITW Chemtronics
C805
15145-450CS 208.44 USD
15145-450
Static Free™ Hand Guard® Lotion, ITW Chemtronics
Skincare Lotions Hand and Body Lotions
This hand lotion is formulated to protect against electrostatic charge
Nongreasy formula contains aloe vera and Vitamin E. Contains no lanolin, silicone, or mineral oil that could inhibit solderability of printed circuits or components. The emollients penetrate deeply to moisten and refresh dry, chapped, or chemically irritated skin.