Static Free™ Mat and Benchtop Reconditioner, ITW Chemtronics
Supplier: ITW Chemtronics
ES1664T
15145-435CS 35.8 USD
15145-435
Static Free™ Mat and Benchtop Reconditioner, ITW Chemtronics
Surface Treatments Floor Treatments
Static-dissipative cleaner and reconditioner for ESD mats, benchtops, and production assembly surfaces can rejuvenate dead spots on ESD mats
This nonflammable, nonabrasive cleaner is noncorrosive and chloride- and amine-free. Safe to use on any surface not affected by water; will not dry out ESD mats.