Cirozane™ Flux-Off® CZ Flux Remover, ITW Chemtronics®
Supplier: ITW Chemtronics
ES7200
15145-426CS 1483.65 USD
15145-426 15145-480
Cirozane™ Flux-Off® CZ Flux Remover, ITW Chemtronics®
Soldering Supplies Flux Removers
Non-ozone depleting, nonflammable flux remover is safe for use on plastics
The BrushClean™ System enables cleaning three ways: brush spot cleaning, flushing under SMDs with the extension tube, and large area cleaning with the spray nozzle. Solvent flow flushes brush clean.
Low surface tension means Flux-Off® CZ flushes tight tolerance areas clean. Cleans Type R, RMA, RA, and most no-clean, synthetic, and water-soluble fluxes. Designed for fast repair and rework on post-solder operations. Evaporates quickly and leaves no residue. Ideal for use on PC boards, SMD pads, sensitive circuit components, component leads, chip carriers, plugs, sockets and heat sinks, thru-hole, and SMT devices. CFC- and HCFC-free.