Flux-Off® Rosin Flux Remover, ITW Chemtronics®
Supplier: ITW Chemtronics
ES1035
15145-442CS 155.81 USD
15145-442 15145-420
Flux-Off® Rosin Flux Remover, ITW Chemtronics®
Soldering Supplies Flux Removers
Plastic-safe, flammable flux remover cleans rosin and encrusted rosin-based no-clean fluxes
The BrushClean™ system enables cleaning three ways: brush spot cleaning, flushing under SMDs with the extension tube, and large area cleaning with the spray nozzle. Solvent flow flushes brush clean.
CFC/HCFC-free and ozone-safe. Dries fast and leaves no residue.