Sample Cups for Chemistry Analyzers, Covidien
Supplier: Cardinal Health
1270016000
15070-057CS 2749.78 USD
15070-057
Sample Cups for Chemistry Analyzers, Covidien
Sample Containers Sample Cups for Analyzers
Samples are poured from blood collection tubes into a cup which is placed inside the tubes, eliminating the need to relabel
Low shoulder height prevents mechanical jamming. Caps are manufactured from high clarity polystyrene. Packaged in polybags.