Hypodermic Syringes with Needles, Terumo®
Supplier: Terumo
SS-03L2138
15003-046CS 217.37 USD
15003-046
Hypodermic Syringes with Needles, Terumo®
Syringes Syringes, Medical
Syringes come with ultra-thin wall, ultra-sharp needles attached
The needles are precisely honed and beveled for ease of penetration. The latex-free clear syringes have bold scale markings allowing quick visual inspection for precise dosage. An integrated plunger stop helps prevent the plunger from being accidentally pulled out