VWR® Stopper Tie
Supplier: VWR International
VSTOPT
15001-372PK 168.81 USD
15001-372
VWR® Stopper Tie
Stoppers Glass Stoppers
Made of stretchable natural rubber for use with glass stoppers.
  • Natural rubber construction
  • Useful for keeping stopcock plug with stored buret
  • Stretchable
Holds stopper to bottle neck to prevent loss when not in use. Stopper tie is also useful for keeping stopcock plug with stored buret. Loop in each end of tie is 9.5 mm I.D. (3/8").