VWR® Signature™ Vacuum Oven Station
Supplier: VWR International
15001-112
15001-112EA 36423.72 USD
15001-112
VWR® Signature™ Vacuum Oven Station
Ovens Vacuum Ovens
VWR® Signature™ Vacuum Oven Station has a programmable Watlow temperature controller, that offers multiple ramp and soak capabilities, including storing and running up to 40 temperature profiles.
  • Large 264 L (9.3 cu. ft.) capacity
  • LED digital control
  • VWR® Two-Year Limited Parts and Labor Warranty
The controls are easily adjustable and the control panel is user-friendly
The oven chamber is wrapped in high temperature insulation which aids overall performance and promotes energy efficiency. Powder coat construction, true vacuum valves and cross-flow ventilation through the oven chamber enhance total performance. The vacuum oven is secured to a ruggedly constructed mobile stand that can store a vacuum pump which saves lab space. Although the oven is not supplied with a vacuum pump, all vacuum plumbing and KF25 connections are provided.
Heat-up time is 1.5 hours at 150 °C. Includes nitrogen purge port, 38 mm (11/2") access port, and auxiliary vacuum port.
A digital vacuum gauge shows chamber vacuum level in measurements of Torr and m/Torr. The display range is 760 Torr down to 0 mm Torr. A secondary independent high limit controller provides overtemperature safety protection.
Ordering information: Supplied with three aluminum shelves. IQ/OQ Validation services are available, contact your Avantor sales representative for further information.