PYREX® Taylor Tube with Lifetime Red Graduations, 50 ml, Corning
Supplier: Corning
7952-25
14852-020CS 57.84 USD
14852-020
PYREX® Taylor Tube with Lifetime Red Graduations, 50 ml, Corning
Tubes Test Tubes
Made of PYREX® borosilicate glass.
  • Reusable
  • Cylindrical in shape
  • Durable white ceramic enamel marking spot on each tube
With beaded rim, designed for general hospital laboratory work. Ideal for determinations involving multiple calibrations and for carrying out digestions. Graduated at 12.5, 25, 35, and 50 ml.
Tolerance: ±0.4 ml