Caraway Capillary Tubes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
42G605 42H606
14705-012CS 1328.44 USD
14705-012 14705-016
Caraway Capillary Tubes, Kimble Chase, DWK Life Sciences
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These Caraway tubes are manufactured with tapered tips for easy collection of micro blood samples.
  • Designed for use with the Caraway technique
  • Packed in lint-free boxes
  • Ends may be sealed with clay or caps
  • Non-heparinized tubes are evenly coated with ammonium heparin to prevent coagulation and do not interfere with sodium identification
Certifications: Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements.
Ordering information: Sodium or lithium heparin is available as a special order.
Packaging: 600/case.