Microhematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
40C505
14705-010CS 258.51 USD
14705-010
Microhematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Heparinized soda lime glass tubes feature a black line at 60mm
Tubes are packed in plastic vials with reclosable snap-lock caps. The ends of the tube may be sealed by flame, clay, or caps.