Micro-Hematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
41B2501 41A2502
14705-003CS 237.8 USD
14705-003 14705-008
Micro-Hematocrit Capillary Tubes, Kimble Chase, DWK Life Sciences
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These Micro-Hematocrit Capillary Tubes are manufactured from soda lime glass, packed in plastic vials with reclosable snap-lock caps to maintain cleanliness.
Ends may be sealed by flame, sealing clay or patented caps. Heparinized capillary tubes are treated to prevent blood clotting.