Oil Sample Bottle, WHEATON®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W216994
14236-000CS 994.66 USD
14236-000
Oil Sample Bottle, WHEATON®, DWK Life Sciences
Bottles Glass Bottles
This bottle is ideal for use as a sample bottle or for storing dry powders, soil samples and specimens.
It is manufactured from Wheaton '800' glass.
Certifications: Meets USP Type III standards.
Ordering information: Cork plug is supplied inserted in bottle. Bulk packed.