SGE Syringe High Pressure Syringe, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
008171
14233-488EA 284.87 USD
14233-488
SGE Syringe High Pressure Syringe, Trajan Scientific and Medical
Syringes Syringes for Chromatography
Designed for high pressure gas sampling (rated to 500 psi).
  • Fitted with a gas tight ON/OFF valve