Uvex® Eyewear Cases, Honeywell Safety
Supplier: Honeywell Safety Products
S487
14233-345EA 2.13 USD
14233-345
Uvex® Eyewear Cases, Honeywell Safety
Safety Glasses
Cases protect lenses against scratches and abrasions.
  • Nylon bag case features a rip-stop drawstring
  • Carry most Uvex® eyewear products
The economy vinyl case has a top closing flap and strap attachment, and holds all flat-fold spectacles. The hard case is equipped with a secure snap closure.
Ordering information: The economy vinyl case is not suitable for Uvex Astro OTG™ 3001 eyewear (see 33000-506 series). The hard case is not suitable for Uvex Astrospec 3000® eyewear (see 33002-036 series).