Super Erecta Shelf® Post-Type and Direct Wall Shelf Mounts, Metro™
Supplier: Metro
SW25C AW21C AW23C AW26C SW31C SW35C SW36C SW43C AW43C AW45C AW46C AW55C AW56C 33PDF 54PDF 13PDFS SBCS 1WS14S 2WS14C 2WS24C 2WS18S 2WS24S 1WD14C 1WD21C 1WD21S 1WD24S 2WD14C 2WD24S SW23C SW26C SW33C AW33C SW45C SW46C SW51C SW56C AW51C 13PDF 63PDF BCS 33PDFS 1WS14C 1WS18C 1WS18S 1WS21S 2WS14S 2WS21S 1WD18S 2WD18C 2WD24C 2WD14S 2WD18S 2WD21S SW21C AW35C SW53C BES 54PDFS 1WS21C 2WS18C 2WS21C 1WD18C 1WD14S AW25C AW31C AW36C SW41C AW41C SW55C AW53C 63PDFS 1WS24C 1WS24S 1WD24C 2WD21C
14233-020EA 592.84 USD
14233-020 14233-024 14233-026 14233-030 14233-032 14233-036 14233-038 14233-050 14233-058 14233-060 14233-062 14233-076 14233-078 14233-082 14233-084 14233-092 14233-102 14233-112 14233-120 14233-126 14233-130 14233-134 14233-136 14233-140 14233-148 14233-150 14233-152 14233-166 14233-018 14233-022 14233-034 14233-042 14233-052 14233-054 14233-064 14233-070 14233-072 14233-080 14233-086 14233-090 14233-094 14233-104 14233-106 14233-114 14233-116 14233-128 14233-132 14233-146 14233-154 14233-158 14233-160 14233-162 14233-164 14233-016 14233-044 14233-066 14233-088 14233-096 14233-108 14233-122 14233-124 14233-138 14233-144 14233-028 14233-040 14233-046 14233-048 14233-056 14233-068 14233-074 14233-098 14233-110 14233-118 14233-142 14233-156
Super Erecta Shelf® Post-Type and Direct Wall Shelf Mounts, Metro™
Furniture Shelves
Wall mounts for shelves are an ideal method for storing items overhead or above work surfaces.
  • All prepackaged post-type units feature a chrome-plated finish
  • Metal endcaps to cover post openings are provided
  • Use single supports at shelf ends and double supports for adjoining shelves
  • Direct wall mounts are available in chrome-plated or stainless steel finish
  • Direct wall systems have a maximum load rating of 91 kg (200 lbs.)
  • Post-type systems have a maximum load rating of 113.3 kg (250 lbs.)
Post-type wall mounts are available in prepackaged end units or mid units, which include all necessary components except wall bolts or screws. End units consist of two posts with brackets, and two single shelf supports for each shelf. Mid units consist of one post with brackets and one double shelf support for each tier. If shelving is to be the length of one shelf, order one end unit only.
Posts, brackets, and shelf supports may also be purchased separately to construct post-type wall mount systems. Each post is supplied with two end brackets. The 85.4 cm (33⁵/₈") long posts also include one intermediate bracket; the 137.2 cm (54") and 157.5 cm (62") long posts also include two intermediate brackets. Intermediate brackets can also be used to attach freestanding shelving to a wall. Post-type components are available in chrome-plated or stainless steel finish.
Direct wall mounts eliminate the need for a post; the shelf support is held by a mounting plate attached directly to the wall. This design also conserves space, as it allows the shelf and shelf support to be much closer to the wall. Each direct wall mount consists of one shelf support, one bracket, and two metal endcaps.
Brackets accommodate all types and sizes of Super Erecta® shelves (shelves sold separately).
Ordering information: If shelving is to be the length of two shelves, order one end unit and one mid unit; if three shelf lengths, order one end and two mid units and so on.