Borosilicate Glass VOA Vials, Sun Sri
Supplier: Thermo Scientific
200 900
14232-706PK 146.19 USD
14232-706 14234-846
Borosilicate Glass VOA Vials, Sun Sri
Vials
VOA vials are made from clear borosillicate glass, and are used for the analysis of volatile organic compounds.
Vials have an operational temperature range from –40 to 125 °C (–40 to 257 °F).
Ordering information: Vials, caps, and septa are available as separate components or as kits.