VWR® Vacuum Oven Gasket
Supplier: VWR International
14231-996
14231-996EA 653.2 USD
14231-996
VWR® Vacuum Oven Gasket
Seals Gaskets
For use with VWR® Vacuum Ovens.
  • Temperature-resistant construction
  • Resilient against wide changes in pressure
  • Fluorosilicone fabrication
Gaskets are designed for specific applications with vacuum ovens.
Max. temperature: 175 °C