Sodium thiosulfate, tablets

Supplier: STARPLEX SCIENTIFIC INC

B1202-TS
14230-838CS 413.73 USD
14230-838
Sodium thiosulfate, tablets
Sodium thiosulphate

This high quality, 120 mL polypropylene container is a terrific product for the environmental market


  • 10 mg Sodium Thiosulfate tablet


It features a white "o-ring" cap, a tamper-proof sterile label and contains a 10 mg Sodium Thiosulfate tablet. It is perfect for analytical water sampling techniques requiring a controlled amount of Sodium Thiosulfate as a dechlorinating agent. By neutralizing free halogens, the tablet prevents destruction of bacteria or organic material prior to analysis. Use for potable water, waste water, recreational waters and surface or irrigation water supplies.

Formula: Na₂S₂O₃
MDL Number: MFCD00003499
CAS Number: 7772-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR