HR2525 Horizontal Gel Electrophoresis System, IBI Scientific
Supplier: IBI Scientific
IB57000
14229-210EA 1194.84 USD
14229-210
HR2525 Horizontal Gel Electrophoresis System, IBI Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
This large-format high throughput system is designed to run up to 200 samples on a single gel in less than 30 minutes
The vented lid reduces condensation, improving output resolution, and reduces heat build-up during electrophoresis, keeping the system running cool. The gravity feed electrode connector eliminates sample disturbance during setup and increases user safety by interrupting power when it is removed. The system is constructed of durable, heavy-duty, 1cm (3/8") thick acrylic. The multichannel pipettor format combs, available with 25 or 50 teeth, reduce the transfer time from plates to gel.
Ideal for running all types of precast gels.
Ordering information: System includes a tank with vented lid, four Delrin® combs, a UV transmissible (UVT) gel casting tray, power cords, a bubble level, and an instruction manual.