Microtiter® 96-Well UV Microplates, Thermo Scientific
Supplier: Thermo Fisher Scientific
8404
14227-680CS 864.37 USD
14227-680
Microtiter® 96-Well UV Microplates, Thermo Scientific
Microplates HTS Microplates
These flat-bottom UV plates offer a low background and higher throughput at wavelengths of 260 and 280nm
Ideal for nucleic acid and protein determinations. Plates are robotic-friendly with a standard 96-well footprint and are perfect for use with any standard UV/Vis spectrophotometer.