Cylindrical Tank, Flat-Bottom, PVDF, Saint-Gobain Performance Plastics
Supplier: Saint Gobain Performance Plastics
11500-0015
14226-870EA 2231.34 USD
14226-870
Cylindrical Tank, Flat-Bottom, PVDF, Saint-Gobain Performance Plastics
Tanks Cylindrical Tanks
Translucent tank with molded-in graduations
  • 2.4 mm (3/32") wall thickness
Constructed of polyvinylidene fluoride (PVDF) that features low extractables, excellent chemical resistance, and inherent UV resistance. Tank has a maximum service temperature of 110°C (230°F). Tank includes a matching cover with the same thickness as the wall.