SolGel-WAX™ GC Columns, SGE
Supplier: Trajan Scientific and Medical
054791 054792 054796 054797 054788 054786 054789 054785 054787 0547100
14226-038EA 1248.46 USD
14226-038 14226-040 14226-048 14226-050 14227-484 14227-486 14227-488 14233-462 14233-466 76409-332
SolGel-WAX™ GC Columns, SGE
Chromatography Columns
Columns feature a phase encapsulated in synthetic glass that is then bonded to the surface of the fused silica.
Temperature range is 30 to 280/300 °C (86 to 536/572 °F). Phase composition is 100% bonded polyethylene glycol. Similar phases are DB®-WAX, Rtx®-Wax, Stabilwax®, HP®-20M, HP®-Wax, HP®-INNOWax, SUPELCOWAX™-10, AT-Wax, Nukol™, CP Wax 52 CB, VB-WAX, and ZB-WAX.