ESD Grid Conductive Sheilding Tape, STATICO
Supplier: Statico
81251
14223-756CS 68.33 USD
14223-756 14223-758 14223-762
ESD Grid Conductive Sheilding Tape, STATICO
Tapes
The S5400 Series is a conductive shielding tape designed for use in static-free workstations, where ESD (Electro Static Discharge) sensitive components and devices are handled
The ESD Grid Conductive Sheilding Tape is supplied on a 3" (72mm) paper core, and is 118' (36m) long.
This tape features anti static inner and outer surfaces, with no shed, flake, crack, chip, or rub-off of the inner layer grids. This combination provides excellent properties for the most effective static prevention and shielding applications.
Certifications: Meets decay rate of MIL-B-81705.