Blood Stain Cards, Qiagen
Supplier: Qiagen
WB100014
14222-822PK 629.64 USD
14222-822
Blood Stain Cards, Qiagen
Nucleic Acid Storage Systems
For the collection and transport of blood and body fluids.
  • Made from absorbent filter paper
  • Enables efficient sample collection
  • Features a protective cover that aids sample drying and protects sample from direct contact
  • Designed for short-term handling of specimens