Loading Dye 6X, IBI Scientific
Supplier: IBI Scientific
IB01010
14221-350PK 86.83 USD
14221-350
Loading Dye 6X, IBI Scientific
Stains Electrophoresis Stains
IBI's 6X Loading Dye contains 15% Ficoll in a special Tris dye, it is ideal for DNA or RNA gels
Dye #1 - Light blue dye is slightly slower than Xylene Cyanol, migrating at around 4 base pairs in a 1% agarose gel.
Dye #2 - Indigo dye migrates similar to Bromophenol Blue, at around 600 base pairs in a 1% agarose gel.
Dye #3 - Magenta dye migrates at around 150 base pairs in a 1% agarose gel.
The 6X Loading Dye contains three tracking dyes that make estimating sample migration easy and reliable.