SMITH & WESSON® MAGNUM 3G™ Safety Glasses KIMBERLY-CLARK PROFESSIONAL®
Supplier: Ansell Healthcare
19822 19940 19794 19961 19799 19823
14219-786EA 19.35 USD
14219-786 14219-788 67200-202 67200-208 14219-760 14219-762 14219-764 14219-774 14219-782 14219-776 14219-778 14219-780 14219-770 14219-772
SMITH & WESSON® MAGNUM 3G™ Safety Glasses KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Safety glasses are ergonomically designed to provide optimal comfort and protection.
  • All lenses provide 99.9% UV protection
  • #3 filter lens coating is ideal for welding sites and for light brazing or cutting
  • #5 filter lens is also ideal for welding sites and for heavy brazing or cutting
Glasses feature a 7-base, cut-to-shape spherical UV blocker lens. Nose pad posts are molded directly onto the lens piece, and retention screws have been replaced with a mechanical snap fit of lens to frame. A built-in hinge eliminates scratching of lens interior. All features aid in ensuring lightweight, secure eye protection. The Mini-Magnum models are equipped with all of the same features on smaller frames for younger wearers.
Certifications: Glasses exceed ANSI Z87.1 standards.