700 System Premium Syringe Barrels and End Caps, Techcon
Supplier: Techcon Systems
703000CRPK 7030LL1DPK 7030LL1NPK 7050LL1BPK 7050LL1DPK 7050LL1NPK 7100LL1BPK 7100LL1DPK 7100LL1NPK 7300LL1BPK 7300LL1DPK
14219-628PK 56.09 USD
14219-628 14219-646 14219-648 14219-652 14219-660 14219-662 14219-666 14219-672 14219-674 14219-678 14219-686 14219-688
700 System Premium Syringe Barrels and End Caps, Techcon
Premium syringe barrels are available in four colors
Natural color is the industry standard. Dark amber color ensures UV and visible light blocking plus the ability to see inside the barrel. Carbon-free black color prevents carbon contamination while providing maximum protection from UV and visible light. Red end caps also block UV light.