700 System Premium Wiper Style Piston
Supplier: TECHCON SYSTEMS INC
Low friction wiper piston can be used with all materials and features a full lip seal. Made of polyethylene. UV blocker. 55,000 cps minimum. Size: 30cc/55cc. Color: white.