700 System Premium Straight Wall Style Piston
Supplier: TECHCON SYSTEMS INC
The air-free loading design of the straight wall piston allows air to bleed while retaining product security. Made of polyethylene. UV blocker. 55,000 cps minimum. Size: 30cc/55cc. Color: red.